Learn More
Abcam Recombinant human Cathepsin S protein (Active), 50UG
Supplier:  Abcam AB28588450UG
Encompass_Preferred
Recombinant human Cathepsin S protein (Active) is a Human Full Length protein, in the 115 to 331 aa range, expressed in Escherichia coli, with >=90% purity and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: Cathepsin S, CTSS
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-221-5598