Learn More
R&D Systems™ Recombinant Human Cbl-B Protein
Description
Specifications
Specifications
| Accession Number | Q13191 |
| For Use With (Application) | Bioactivity |
| Formulation | X mg/ml (X μM) in 50 mM HEPES pH 7.5, 200 mM NaCl, 10% (v/v) Glycerol, 2 mM DTT |
| Gene ID (Entrez) | 868 |
| Molecular Weight (g/mol) | M.W.-theoretical: 110 kDa |
| Quantity | 50 μg |
| Source | Spodoptera frugiperda, Sf 21 (baculovirus)-derived human Cbl-B protein |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 48 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Gene Alias | Cas-Br-M (murine) ecotropic retroviral transforming sequence b, Cas-Br-M (murine) ectropic retroviral transforming sequence b, Casitas B-lineage lymphoma proto-oncogene b, CBLB, DKFZp686J10223, DKFZp779A0729, DKFZp779F1443, E3 ubiquitin-protein ligase CBL-B, EC 6.3.2, EC 6.3.2.-, FLJ41152, Nbla00127, RING finger protein 56, RNF56, RNF56FLJ36865, SH3-binding protein CBL-B, Signal transduction protein CBL-B |
| Conjugate | Unconjugated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.