Learn More
Biomatik Corporation Recombinant Human C-C chemokine receptor type 8 (CCR8)
Supplier:  Biomatik Corporation RPC27433
Encompass_Preferred
Recombinant Human C-C chemokine receptor type 8 (CCR8) is a purified CF Transmembrane Protein. Purity: >85% as determined by SDS-PAGE. Host: In Vitro E. coli Expression System. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: CCR8. Target Synonyms: CC chemokine receptor CHEMR1CMKBRL2Chemokine receptor-like 1CKR-L1GPR-CY6GPRCY6TER1CD_antigen: CDw198. Accession Number: P51685. Expression Region: 1~355aa. Tag Info: C-Terminal 6Xhis-Tagged. Theoretical MW: 41.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7922