Learn More
Biomatik Corporation Recombinant Human C-C chemokine receptor type 8 (CCR8)-VLPs, Active Protein
Supplier:  Biomatik Corporation RPC28100
Encompass_Preferred
Recombinant Human C-C chemokine receptor type 8 (CCR8)-VLPs, Active Protein is a purified MP-VLP Transmembrane Protein; Active Protein. Purity: Testing in progress. Host: Mammalian Cell. Endotoxin Level: <1.0 EU/ug as determined by LAL method. Species: Human (Homo sapiens). Target Name: CCR8. Target Synonyms: CC chemokine receptor CHEMR1; CMKBRL2; Chemokine receptor-like 1; CKR-L1; GPR-CY6; GPRCY6; TER1; CDw198; CKRL1; CMKBR8; CMKBRL2. Accession Number: P51685. Expression Region: 1~355aa. Tag Info: C-Terminal 10Xhis-Tagged (This Tag Can Be Tested Only Under Denaturing Conditions). Theoretical MW: 42.2kDa Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6180