Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human CCL3/MIP-1 alpha Protein

Catalog No. p-4719261
Change view
Click to view available options
Quantity:
10 μg
50 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
270LD010 10 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
270LD050CF 50 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
270LD010CF 10 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
270LD050 50 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 270LD010 Supplier R&D Systems™ Supplier No. 270LD010
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human CCL3/MIP-1 alpha Protein is derived from E. coli. The Recombinant Human CCL3/MIP-1 alpha Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Gene ID (Entrez) 6348
Molecular Weight (g/mol) 7.5 kDa
Name CCL3/MIP-1 alpha Isoform LD78a
Quantity 10 μg
Source E. coli-derived human CCL3/MIP-1 alpha protein Ala27-Ala92
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Gene Alias C-C motif chemokine 3, MIP1-(a), AI323804, chemokine (C-C motif) ligand 3, G0S19-1, LD78a, LD78alpha, MIP1 alpha, MIP-1 alpha, Mip1a, MIP-1alpha, MIP1-alpha, PAT 464.1, SCYA3, SIS-beta
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.