Learn More
Abcam Recombinant Human CD133 protein, 10UG
Supplier:  Abcam AB16021810UG
Encompass_Preferred
Recombinant Human CD133 protein is a Human Full Length protein, in the 1 to 856 aa range, expressed in Wheat germ and suitable for SDS-PAGE, ELISA, WB.
Protein/ Peptide/ Enzyme Synonyms: CD133, PROML1, MSTP061, PROM1, Prominin-1, Antigen AC133, Prominin-like protein 1
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1903561