Learn More
R&D Systems™ Recombinant Human CD160 Fc Chimera Protein, CF
Specifications
Specifications
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 11126 |
| Molecular Weight (g/mol) | M.W.-observed: 39-59 kDa, under reducing conditions, M.W.-theroretical: 41 kDa |
| Quantity | 50 μg |
| Storage Requirements | Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Gene Alias | BY55, BY55FLJ46513, CD160, CD160 antigenimmunoglobulin superfamily member, CD160 molecule, Natural killer cell receptor BY55, NK1, NK28 |
| Conjugate | Unconjugated |
| Reconstitution | Reconstitute at 250μg/mL in sterile water. |
| Protein | CD160 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.