Learn More
Medchemexpress LLC Recombinant human CD1b protein (HEK293, C-hFc) | >95.0% | 10 UG
SDP
Catalog No. 5000293419
Encompass_Preferred
Recombinant human CD1b protein expressed in HEK293 mammalian cells with a C-terminal Fc tag, provided as a research-grade recombinant protein suitable for immunology, antigen presentation, and assay development applications. Supplied in small pack sizes for laboratory use.
  • Expressed in HEK293 mammalian cells with C-terminal Fc tag
  • Reported purity >95% as determined by reducing SDS-PAGE
  • Sequence region reported as S18-S303 (UniProt accession P29016)
  • Stable at -20 °C; after reconstitution stable at 4 °C for 1 week or -20 °C for longer with carrier protein
  • Available in small research pack sizes suitable for assay development and characterization
Catalog No. 50-002-93419 Supplier Medchemexpress LLC Supplier No. HYP7543810UG
Recombinant human CD1b protein expressed in HEK293 mammalian cells with a C-terminal Fc tag, provided as a research-grade recombinant protein suitable for immunology, antigen presentation, and assay development applications. Supplied in small pack sizes for laboratory use.
  • Expressed in HEK293 mammalian cells with C-terminal Fc tag
  • Reported purity >95% as determined by reducing SDS-PAGE
  • Sequence region reported as S18-S303 (UniProt accession P29016)
  • Stable at -20 °C; after reconstitution stable at 4 °C for 1 week or -20 °C for longer with carrier protein
  • Available in small research pack sizes suitable for assay development and characterization