Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human CD27 Ligand/TNFSF7 Protein

Catalog No. 9328CL100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
93-28C-L100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 93-28C-L100 Supplier R&D Systems™ Supplier No. 9328CL100
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to induce IL-8 secretion in HT1080 human fibrosarcoma cells transfected with human CD27. The ED50 for this effect is 5-25 ng/mL in the presence of Mouse Anti-Hemagglutinin/HA Peptide Monoclonal Antibody (Catalog # MAB060 ).

Order Info

Ships at ambient temperature.

Specifications

Accession Number P32970
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 970
Molecular Weight (g/mol) M.W. (Observed): 24-32 kDa, reducing conditions; M.W. (theoretical): 23 kDa
Quantity 100 μg
Source Human embryonic kidney cell, HEK293-derived human CD27 Ligand/TNFSF7 protein HA (YPYDVPDYA) GCN4-IZ GGGSGGGSGGGS Human CD27 Ligand (Gln39-Pro193) (N-terminus) Accession # P32970 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Regulatory Status RUO
Structural Form Noncovalently-linked homotrimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Biological Activity 5 to 25ng/mL
Product Type Recombinant protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Reconstitute at 200 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein CD27 Ligand/TNFSF7/CD70
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.