Learn More
Creative Biomart Recombinant Human CD27 protein, Fc-tag, GMP Grade
Supplier:  Creative Biomart CD27079THP
Encompass_Preferred
Recombinant Human CD27 protein GMP Grade(NP_001233.1)(394 aa) was expressed in HEK293 cells with a C-terminal Fc Tag.
Catalog No. 50-208-5319