Learn More
Creative Biomart Recombinant Human CD274 protein, His-tag, GMP Grade
Supplier:  Creative Biomart CD274702THP
Encompass_Preferred
Recombinant Human CD274 protein GMP Grade(NP_054862.1)(222 aa) was expressed in HEK293 cells with a C-terminal His Tag.
Catalog No. 50-208-5329