Learn More
Medchemexpress LLC Recombinant human CD299 (CLEC4M) Fc-fusion protein | 90.1% | 50 UG
SDP
Catalog No. 5000290725
Encompass_Preferred
Recombinant human CD299 extracellular domain fused to human Fc, expressed in HEK293 cells and supplied as a lyophilized powder. Designed for binding assays, immunoassays, and protein-protein interaction studies, the product has low endotoxin and is supplied in a PBS-based formulation for reliable reconstitution and downstream functional assays.
  • Expressed in HEK293 cells for proper mammalian glycosylation
  • Human Fc fusion enhances stability and facilitates detection
  • Lyophilized powder formulated in PBS, pH 7.4, for storage stability
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays
  • Recommended reconstitution ≥100 μg/mL for functional use
  • Contains extracellular sequence S78-E399 of the human protein
Catalog No. 50-002-90725 Supplier Medchemexpress LLC Supplier No. HYP76219A50UG
Recombinant human CD299 extracellular domain fused to human Fc, expressed in HEK293 cells and supplied as a lyophilized powder. Designed for binding assays, immunoassays, and protein-protein interaction studies, the product has low endotoxin and is supplied in a PBS-based formulation for reliable reconstitution and downstream functional assays.
  • Expressed in HEK293 cells for proper mammalian glycosylation
  • Human Fc fusion enhances stability and facilitates detection
  • Lyophilized powder formulated in PBS, pH 7.4, for storage stability
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays
  • Recommended reconstitution ≥100 μg/mL for functional use
  • Contains extracellular sequence S78-E399 of the human protein