Learn More
R&D Systems™ Recombinant Human CD3 epsilon Fc Chimera Protein
Description
Specifications
Specifications
| Accession Number | P07766 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 916 |
| Molecular Weight (g/mol) | 38 kDa |
| Name | CD3e, CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, FLJ18683, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell,HEK293-derived human CD3 epsilon protein, (N-terminus) Human CD3 epsilon (Gln22-Asp126) Accession &Num; P07766 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | 12 months from date of receipt, ≤ -20° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, ≤ -20° C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1μg of the protein by the LAL method. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.