Learn More
Abcam Recombinant human CD3 epsilon protein (Active), 100UG
Supplier:  Abcam AB167755100UG
Encompass_Preferred
Recombinant human CD3 epsilon protein (Active) is a Human Fragment protein, in the 23 to 126 aa range, expressed in HEK 293, with >90% purity, < 1 EU/µg endotoxin level and suitable for SDS-PAGE, FuncS.
Protein/ Peptide/ Enzyme Synonyms: CD3e, T3E, CD3E, T-cell surface glycoprotein CD3 epsilon chain, T-cell surface antigen T3/Leu-4 epsilon chain
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1473647