Learn More
R&D Systems™ Recombinant Human CD300e Fc Chimera Alexa Fluor™ 488 Protein
Description
Specifications
Specifications
| Accession Number | Q496F6.2 |
| For Use With (Application) | Bioactivity |
| Formulation | Supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 342510 |
| Molecular Weight (g/mol) | M.W.-observed: 45-58 kDa and 106-125 kDa (non-reducible dimer), under reducing conditions., M.W.-theroretical: 44 kDa |
| Quantity | 20 μg |
| Source | Human embryonic kidney cell, HEK293-derived human CD300e/LMIR6 protein Human CD300e/LMIR6 (Leu18-Leu169) (N-terminus) Accession # Q496F6.2 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | CD300 antigen like family member E, CD300 antigen-like family member E, CD300e, CD300e antigenPIgR-2, CD300e molecule, CD300LE, CD300LEPIgR2, CLM2, CLM-2, CLM2CMRF35-A5, CMRF35A5, CMRF35-like molecule 2, Immune receptor expressed on myeloid cells 2, IREM-2, IREM2IREM-2, LMIR6, Poly-Ig receptor 2, Polymeric immunoglobulin receptor 2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.