Learn More
R&D Systems™ Recombinant Human CD40/TNFRSF5 Fc Alexa Fluor™ 488 Protein
Description
Specifications
Specifications
| Accession Number | P25942-1 |
| For Use With (Application) | Bioactivity |
| Formulation | Supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 958 |
| Molecular Weight (g/mol) | M.W.-observed: 58-66 kDa, under reducing conditions., M.W.-theroretical: 47 kDa |
| Quantity | 20 μg |
| Source | Mouse myeloma cell line, NS0-derived human CD40/TNFRSF5 protein Human CD40 (Glu21-Arg193) (C-terminus) Accession # P25942-1 DIEGRMD Human IgG1 (Pro100-Lys330) 6-His tag (N-terminus) |
| Storage Requirements | Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | B cell surface antigen CD40, B-cell surface antigen CD40, Bp50B cell-associated molecule, CD40, CD40 antigen, CD40 molecule, TNF receptor superfamily member 5, CD40 type II isoform, CD40L receptor, CDw40, MGC9013, nerve growth factor receptor-related B-lymphocyte activation molecule, p50, TNFRSF5, TNFRSF5CD40 antigen (TNF receptor superfamily member 5), tumor necrosis factor receptor superfamily member 5, tumor necrosis factor receptor superfamily, member 5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.