Learn More
R&D Systems™ Recombinant Human CD42b/GPIb alpha Protein
Description
Specifications
Specifications
| Accession Number | P07359.1 |
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 2811 |
| Molecular Weight (g/mol) | 54.7 kDa |
| Name | CD42b/GPIb alpha |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived human CD42b/GPIb alpha protein His17-Leu505,with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.1 EU per 1μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.