Learn More
Creative Biomart Recombinant Human CD47 protein, His-tag, GMP Grade
Catalog No. 502085324
Encompass_Preferred
Recombinant Human CD47 protein GMP Grade(NP_942088.1)(121 aa) was expressed in HEK293 cells with a C-terminal His Tag.
Catalog No. 50-208-5324 Supplier Creative Biomart Supplier No. CD47101THP
Add to Cart
Add to Cart
Recombinant Human CD47 protein GMP Grade(NP_942088.1)(121 aa) was expressed in HEK293 cells with a C-terminal His Tag.