Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human CD52 Fc Chimera Protein

Catalog No. 9116CD050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
9116CD050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 9116CD050 Supplier R&D Systems™ Supplier No. 9116CD050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human CD52 Fc Chimera Protein is derived from HEK293. The Recombinant Human CD52 Fc Chimera Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number P31358
For Use With (Application) Bioactivity
Gene ID (Entrez) 1043
Molecular Weight (g/mol) Observed M.W.: 38 - 44kDa, reducing conditions, Theoretical M.W.: 28kDa
Quantity 50 μg
Source Human embryonic kidney cell,HEK293-derived human CD52 protein, (N-terminus) Human CD52 (Gly25-Ser36) Accession &Num; P31358 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias Cambridge pathology 1 antigen, CAMPATH-1, CAMPATH-1 antigen, CD52, CD52 antigen, CD52 antigen (CAMPATH-1 antigen), CD52 molecule, CDw52, CDW52 antigen (CAMPATH-1 antigen), Epididymal secretory protein E5, HE5, HEL-S-171mP, Human epididymis-specific protein 5
Common Name CD52
Gene Symbol CD52
Biological Activity Measured by its binding ability in a functional ELISA. When Recombinant Human CD52 Fc Chimera is immobilized at 1μg/mL, 100μL/well, the concentration of Recombinant Human Siglec-10 Fc Chimera that produces 50% of the optimal binding response is approximately 1-5μg/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Human embryonic kidney cell, HEK293-derived Human CD52 (Gly25-Ser36) Accession # P31358, N-terminus; IEGRMD; Human IgG1 (Pro100-Lys330), C-terminus
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.