Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human CD68/SR-D1 Fc Chimera Protein

Catalog No. 10301SR050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-301-SR050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-301-SR050 Supplier R&D Systems™ Supplier No. 10301SR050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Measured by its binding ability in a functional ELISA.

When Recombinant Human S100A8 is immobilized at 10 μg/mL (100 μL/well), it binds to Recombinant Human CD68/SR-D1 Fc Chimera with an ED50 of 5 to 30 μg/mL

Specifications

Accession Number P34810
For Use With (Application) Bioactivity
Gene ID (Entrez) 968
Molecular Weight (g/mol) 58 kDa
Name CD68, CD68 antigenmacrophage antigen CD68, CD68 molecule, DKFZp686M18236, gp110, Macrosialin, SCARD1, scavenger receptor class D, member 1, SRD1, SR-D1
Quantity 50 μg
Source

Mouse myeloma cell line, NS0-derived human CD68/SR-D1 protein

Human CD68/SR-D1 (Asn22-Ile320) Accession # P34810IEGRMDHuman IgG1 (Pro100-Lys330)N-terminus C-terminus

Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 500 μg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS.
Endotoxin Level <0.10 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.