Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human CD74 Protein, CF

Catalog No. 3590CDB050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-90C-DB050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 35-90C-DB050 Supplier R&D Systems™ Supplier No. 3590CDB050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Measured by its binding ability in a functional ELISA. Recombinant Human CD74 (Catalog # 3590-CDB) binds to Human CD74 Antibody (Catalog # AF3590 ) with an ED50 of 0.300-3.60 ng/mL.

Specifications

Accession Number NP_004346.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 972
Molecular Weight (g/mol) M.W.-observed: 25-33 kDa, under reducing conditions., M.W.-theroretical: 19 kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived human CD74 protein Gln73-Met232, with an N-terminal HA (YPYDVPDYA) tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias CD74, CD74 antigen, CD74 antigen (invariant polypeptide of major histocompatibility complex, classII antigen-associated), CD74 molecule, major histocompatibility complex, class II invariant chain, CLIP, DHLAG, DHLAGgamma chain of class II antigens, HLA class II histocompatibility antigen gamma chain, HLADG, HLA-DR antigens-associated invariant chain, HLA-DR-gamma, Ia antigen-associated invariant chain, Ia-associated invariant chain, Ia-GAMMA, Ii, INVG34, MHC HLA-DR gamma chain, p33
Conjugate Unconjugated
Reconstitution Reconstitute at 250 μg/mL in water.
Protein CD74
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.