Learn More
R&D Systems™ Recombinant Human CD94 Fc Chimera Protein
Description
Specifications
Specifications
| Accession Number | Q13241.2 |
| For Use With (Application) | Bioactivity |
| Formulation | Supplied as a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 3824 |
| Molecular Weight (g/mol) | M.W.-theroretical: 44 kDa, M.W.-observed: 53-62 kDa, under reducing conditions |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived human CD94 protein MD Human IgG1 (Pro100-Lys330) IEGR Human CD94 (Lys32-Ile179) (c-terminus) Accession # Q13241.2 N-terminus C-terminμs |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | CD94, CD94 antigen, CD94natural killer cells antigen CD94, Killer cell lectin-like receptor subfamily D member 1, killer cell lectin-like receptor subfamily D, member 1, KLRD1, KP43, NK cell receptor |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.