Learn More
Abcam Recombinant Human CECR1 protein (His tag), 50UG
Supplier:  Abcam AB21921750UG
Encompass_Preferred
Recombinant Human CECR1 protein (His tag) is a Human Full Length protein, in the 30 to 511 aa range, expressed in Baculovirus infected insect, with >90% purity, < 1 EU/µg endotoxin level and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: ADGF, CECR1, IDGFL, ADA2, Adenosine deaminase 2, Cat eye syndrome critical region protein 1
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2541056