Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Recombinant human cell adhesion molecule 2 (CADM2) | >95.0% | 10 UG
SDP

Catalog No. 5000291795
Encompass_Preferred

Recombinant human cell adhesion molecule 2 (CADM2/IGSF4D) extracellular region (partial) produced in HEK293 cells and supplied as a His-tagged, lyophilized protein for research use. The protein is glycosylated, has an apparent molecular weight of ~40-57 kDa, and is provided with high purity and low endotoxin. Lyophilized from PBS (pH 7.4); store at -20°C and reconstitute to ≥100 μg/mL in ddH2O with carrier protein for long-term storage.

  • Expression system: HEK293 cells.
  • Tag: C-terminal 6xHis.
  • Sequence: residues G30-D335 (partial).
  • Molecular weight: ~40-57 kDa (glycosylated).
  • Purity: >95.0% by reducing SDS-PAGE.
  • Endotoxin: <1 EU/μg (LAL).
  • Formulation: lyophilized from PBS, pH 7.4.
  • Appearance: lyophilized powder.
  • Storage: store at -20°C; aliquot to avoid repeated freeze-thaw.

Catalog No. 50-002-91795 Supplier Medchemexpress LLC Supplier No. HYP7559910UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human cell adhesion molecule 2 (CADM2/IGSF4D) extracellular region (partial) produced in HEK293 cells and supplied as a His-tagged, lyophilized protein for research use. The protein is glycosylated, has an apparent molecular weight of ~40-57 kDa, and is provided with high purity and low endotoxin. Lyophilized from PBS (pH 7.4); store at -20°C and reconstitute to ≥100 μg/mL in ddH2O with carrier protein for long-term storage.

  • Expression system: HEK293 cells.
  • Tag: C-terminal 6xHis.
  • Sequence: residues G30-D335 (partial).
  • Molecular weight: ~40-57 kDa (glycosylated).
  • Purity: >95.0% by reducing SDS-PAGE.
  • Endotoxin: <1 EU/μg (LAL).
  • Formulation: lyophilized from PBS, pH 7.4.
  • Appearance: lyophilized powder.
  • Storage: store at -20°C; aliquot to avoid repeated freeze-thaw.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.