Learn More
Abcam Recombinant Human CFTR protein, 10UG
Supplier:  Abcam AB11424610UG
Encompass_Preferred
Recombinant Human CFTR protein is a Human Fragment protein, in the 1381 to 1480 aa range, expressed in Wheat germ and suitable for SDS-PAGE, ELISA, WB.
Protein/ Peptide/ Enzyme Synonyms: ABCC7, CFTR, Cystic fibrosis transmembrane conductance regulator, ATP-binding cassette sub-family C member 7, Channel conductance-controlling ATPase, cAMP-dependent chloride channel
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2172424