Learn More
R&D Systems™ Recombinant Human CLEC14A Fc Chimera Protein
Description
Specifications
Specifications
| Accession Number | Q86T13 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 161198 |
| Molecular Weight (g/mol) | 66 kDa |
| Name | C14orf27, CEG1, chromosome 14 open reading frame 27, ClECT and EGF-like domain containing protein, C-type lectin domain family 14 member A, C-type lectin domain family 14, member A, EGF R5, EGFR5, EGFR-5, Epidermal growth factor receptor 5 |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell,HEK293-derived human CLEC14A protein, (N-terminus) Human CLEC14A (Glu22-Ala397) Accession &Num; Q86T13 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1μg of the protein by the LAL method. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.