Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human CNTF Protein

Catalog No. p-4719365
Change view
Click to view available options
Quantity:
10 μg
50 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS, NaCl, EDTA, DTT and CHAPS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS, NaCl, EDTA, DTT and CHAPS.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
257NT010 10 μg Lyophilized from a 0.2μm filtered solution in PBS, NaCl, EDTA, DTT and CHAPS with BSA as a carrier protein.
257NT050 50 μg Lyophilized from a 0.2μm filtered solution in PBS, NaCl, EDTA, DTT and CHAPS with BSA as a carrier protein.
257NT010CF 10 μg Lyophilized from a 0.2μm filtered solution in PBS, NaCl, EDTA, DTT and CHAPS.
257NT050CF 50 μg Lyophilized from a 0.2μm filtered solution in PBS, NaCl, EDTA, DTT and CHAPS.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 257NT010 Supplier R&D Systems™ Supplier No. 257NT010
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human CNTF Protein is derived from E. coli. The Recombinant Human CNTF Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS, NaCl, EDTA, DTT and CHAPS with BSA as a carrier protein.
Gene ID (Entrez) 1270
Molecular Weight (g/mol) 22.8 kDa
Name CNTF
Quantity 10 μg
Source E. coli-derived human CNTF protein Ala2-Met200
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 3 months, 2 to 8° C under sterile conditions after reconstitution.
Structural Form This recombinant protein is prone to proteolytic degradation at the C-terminus, resulting in proteins with molecular masses of 22.2 - 22.8 kDa.
Endotoxin Concentration <0.1 EU per 1μg of the protein by the LAL method.
Conjugate Unconjugated
Recombinant Recombinant
Activity Measured in a cell proliferation assay using TF-1 human erythroleukemic cells. Kitamura, T. et al. (1989) J. Cell Physiol. 140:323. The ED50 for this effect is 0.03-0.18 μg/mL. The proliferative effect can be enhanced up to 100-fold by the addition of Recombinant Human CNTF R alpha (Catalog # 303-CR ) .
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.