Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Collagen IV alpha 1/Arresten Protein, CF

Catalog No. 11597CL050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-597-CL050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-597-CL050 Supplier R&D Systems™ Supplier No. 11597CL050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

His-tag

Measured by its binding ability in a functional ELISA. Recombinant Human Collagen IV alpha 1 His-tag (Catalog # 11597-CL) binds Recombinant Human Integrin alpha 1 beta 1 (Catalog # 7064-AB ) with an ED50 of 85.0-850 ng/mL.

Specifications

Accession Number P02462.3
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 1282
Molecular Weight (g/mol) M.W.-observed: 22-33 kDa, under reducing conditions, M.W.-theroretical: 26 kDa
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived human Collagen IV alpha 1 protein Ser1441-Thr1669, with a C-ter 6xHis tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias alpha-1 chain, arresten, COL4A1, COL4A1 NC1 domain, collagen alpha-1(IV) chain, collagen IV, alpha-1 polypeptide, collagen, type IV, alpha 1, Collagen-4, EC 3.4.23, EC 6.3.1.2
Conjugate Unconjugated
Reconstitution Reconstitute at 250μg/mL in PBS.
Protein Collagen IV alpha 1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.