Learn More
Biomatik Corporation Recombinant Human coronavirus OC43 Spike glycoprotein (S), partial
Supplier:  Biomatik Corporation RPC22573
Encompass_Preferred
Recombinant Human coronavirus OC43 Spike glycoprotein (S), partial is a purified Recombinant Protein; Coronavirus Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human coronavirus OC43 (HCoV-OC43). Target Name: S. Target Synonyms: S; 3Spike glycoprotein; S glycoprotein; E2; Peplomer protein; Cleaved into: Spike protein S1; Spike protein S2; Spike protein S2'. Accession Number: P36334. Expression Region: 15~344aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 53.6kDa. Buffer: Lyophilized from Tris/PBS-based buffer, 6% Trehalose, pH 8.0 Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7280