Learn More
R&D Systems™ Recombinant Human CUL1/RBX1 Neddylated Complex Protein, CF
Description
Specifications
Specifications
| Accession Number | Q13616.2 (CUL1), P62877.1 (RBX1) |
| Formulation | Supplied as a 0.2 μm filtered solution in HEPES, NaCl, DTT and Glycerol. |
| Molecular Weight (g/mol) | M.W. Theroretical: 104 kDa (neddylated CUL1), 12 kDa (RB x 1) |
| Quantity | 25 μg |
| Source | Spodoptera frugiperda, Sf 21 (baculovirus)-derived human CUL1/RBX1 Complex protein Met1 - Ala776 with a N-terminal 10-His Sumo tag (CUL1), Met1 - His108 (RBX1) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Purity or Quality Grade | >90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie Blue stain. |
| Protein | CUL1/RBX1 Complex |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.