Learn More
R&D Systems™ Recombinant Human CUL2/RBX1 Complex Protein
Description
Specifications
Specifications
| Accession Number | Q13617.2 (CUL2), P62877.1 (RBX1) |
| For Use With (Application) | Bioactivity |
| Formulation | Supplied as a 0.2 μm filtered solution in HEPES, NaCl, DTT and Glycerol. |
| Molecular Weight (g/mol) | M.W. (theoretical): 93 kDa (CUL2), 12 kDa (RB x 1) |
| Quantity | 25 μg |
| Source | Spodoptera frugiperda, Sf 21 (baculovirus)-derived human CUL2/RB x 1 Complex protein Met1 - Ala 745 with a N-terminal 10-His Sumo tag (CUL2), Met1 - His108 (RB x 1) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Species | Human |
| Buffer | X mg/mL (X μm) in 50 mm HEPES pH 7.5, 200 mm NaCl, 10% (v/v) Glycerol, 1 mm DTT |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.