Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Cyclophilin A Protein, CF

Catalog No. 3589CAB100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-89C-AB100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 35-89C-AB100 Supplier R&D Systems™ Supplier No. 3589CAB100
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

The Recombinant Human Cyclophilin A Protein is derived from E. coli. The Recombinant Human Cyclophilin A Protein has been validated for the following applications: Enzyme Activity.

Specifications

Accession Number P62937
For Use With (Application) Enzyme Activity
Gene ID (Entrez) 5478
Molecular Weight (g/mol) 18 kDa
Quantity 100 μg
Source E. coli-derived human Cyclophilin A protein Met1-Glu165
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70 °C as supplied. 3 months, -20 to -70 °C under sterile conditions after opening.
Regulatory Status RUO
Endotoxin Concentration <1.0 EU per 1 μg of the protein by the LAL method.
Specific Reactivity Cyclophilin A
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Buffer Supplied as a 0.2 μm filtered solution in MES and NaCl.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie« Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.