Learn More
R&D Systems™ Recombinant Human DC-SIGNR/CD299 Fc Chimera Protein
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in Tris-Citrate. |
| Gene ID (Entrez) | 10332 |
| Molecular Weight (g/mol) | 61.4 kDa (monomer) |
| Name | DC-SIGNR/CD299 Fc Chimera |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived human DC-SIGNR/CD299 protein, (N-terminus) MD Human IgG1 (Pro100-Lys330) IEGR Human DC-SIGNR (Ser73-Glu376) Accession &Num; NP_001138382 (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked homodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.