Learn More
Biomatik Corporation Recombinant Human Deoxyribonuclease gamma (DNASE1L3)
Supplier:  Biomatik Corporation RPC26147
Encompass_Preferred
Recombinant Human Deoxyribonuclease gamma (DNASE1L3) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: DNASE1L3. Target Synonyms: DNase I homolog protein DHP2 (Deoxyribonuclease I-like 3; DNase I-like 3; Liver and spleen DNase; LS-DNase; LSD; DNase gamma; DHP2; DNAS1L3). Accession Number: Q13609. Expression Region: 21~305aa. Tag Info: Tag-Free. Theoretical MW: 33.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7930