Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human DHFR Protein

Catalog No. 8456DR100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
84-56D-R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 84-56D-R100 Supplier R&D Systems™ Supplier No. 8456DR100
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human DHFR Protein is derived from E. coli. The Recombinant Human DHFR Protein has been validated for the following applications: Enzyme Activity.

Specifications

Accession Number P00374
For Use With (Application) Enzyme Activity
Gene ID (Entrez) 1719
Molecular Weight (g/mol) Observed M.W.: 23kDa, reducing conditions, Theoretical M.W.: 22kDa
Quantity 100 μg
Source Mouse myeloma cell line, NS0-derived human CD30/TNFRSF8 protein Human CD30 (Phe19-Lys379) (N-terminus) Accession # P28908.1 IEGRDMD Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU per 1μg of the protein by the LAL method.
Gene Alias DHFR, DHFRP1, dihydrofolate reductase, DYR, EC 1.5.1.3
Common Name Dihydrofolate Reductase/DHFR
Gene Symbol DHFR
Biological Activity Measured by the reduction of dihydrofolic acid (DHF). The specific activity is >5,500 pmol/min/μg, as measured under the described conditions.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System E. coli-derived Met1-Asp187, with a C-terminal 6-His tag
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.