Learn More
Biomatik Corporation Recombinant Human E3 ubiquitin-protein ligase TRIM21 (TRIM21)
Supplier:  Biomatik Corporation RPC22174
Encompass_Preferred
Recombinant Human E3 ubiquitin-protein ligase TRIM21 (TRIM21) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: TRIM21. Target Synonyms: 52 kDa Ro protein52 kDa ribonucleoprotein autoantigen Ro/SS-ARING finger protein 81RING-type E3 ubiquitin transferase TRIM21Ro(SS-A)Sjoegren syndrome type A antigenSS-ATripartite motif-containing protein 21RNF81, RO52, SSA1. Accession Number: P19474. Expression Region: 1~475aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 58.2kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5540