Learn More
R&D Systems™ Recombinant Human EGFR Fc Chimera Alexa Fluor™ 488 Protein
Description
Specifications
Specifications
| Accession Number | AAH17865.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein. |
| Molecular Weight (g/mol) | M.W.-observed: 40-50 kDa, under reducing conditions.; M.W.-theoretical: 22.6 kDa |
| Quantity | 20 μg |
| Source | Mouse myeloma cell line, NS0-derived human Fc gamma RIIIA/CD16a protein Gly17-Gln208, with a C-terminal 6-His tag |
| Storage Requirements | Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after opening. 3 months, -20 to -70 °C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU per 1 μg of the protein by the LAL method. |
| Conjugate | Alexa Fluor 488 |
| Purity or Quality Grade | >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.