Learn More
Creative Biomart Recombinant Human EGFR protein, His-tag, GMP Grade
Supplier:  Creative Biomart EGFR078THP
Encompass_Preferred
Recombinant Human EGFR protein GMP Grade(NP_005219)(620 aa) was expressed in HEK293 cells with a C-terminal His Tag.
Catalog No. 50-208-5298