Learn More
Biomatik Corporation Recombinant Human Egl nine homolog 1 (EGLN1), partial
Supplier:  Biomatik Corporation RPC28302
Encompass_Preferred
Recombinant Human Egl nine homolog 1 (EGLN1), partial is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: EGLN1. Target Synonyms: Hypoxia-inducible factor prolyl hydroxylase 2 (HIF-PH2; HIF-prolyl hydroxylase 2; HPH-2; Prolyl hydroxylase domain-containing protein 2; PHD2; SM-20; C1orf12; PNAS-118; PNAS-137). Accession Number: Q9GZT9. Expression Region: 177~426aa. Tag Info: N-Terminal 10Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 35.3kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7236