Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Embigin/EMB Fc Chimera Protein CF

Catalog No. 10832EM050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10832EM050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10832EM050 Supplier R&D Systems™ Supplier No. 10832EM050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Measured by its ability to inhibit neurite outgrowth of E16-E18 rat embryonic cortical neurons. Recombinant Human Embigin/EMB Fc Chimera, immobilized at 2.5 μg/mL on a 96 well plate, is able to significantly inhibit neurite outgrowth.

Specifications

Accession Number Q6PCB8.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 133418
Molecular Weight (g/mol) M.W.-observed: 85-100 kDa, under reducing conditions, M.W.-theroretical: 52 kDa
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived human Embigin/EMB protein Human Embigin/EMB (Asp33-Pro260) (N-terminus) Accession # Q6PCB8.1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias EMB, embigin, embigin homolog, embigin homolog (mouse), F9, GP70, H, MGC71745
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Embigin homolog
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.