Learn More
Proteintech Group Inc Recombinant Human EPO 10ug
SDP
Supplier:  Proteintech Group Inc HZ116810UG
Encompass_Preferred
Animal-free Recombinant Human EPO is expressed in human 293 cells in a serum-free, chemically defined media. EPO has more abundant and diverse glycan profiles and exhibits a lower molecular mass and substantially higher content of neutral glycans than the CHO cell produced version. See “EPO Authentic Glycan Structure” article for more information on the comparison of glycan structure.
Catalog No. 50-197-3922