Learn More
Creative Biomart Recombinant Human EPO protein, His-tag, GMP Grade
Supplier:  Creative Biomart EPO323THP
Encompass_Preferred
Recombinant Human EPO protein GMP Grade(NP_000790.2)(172 aa) was expressed in HEK293 cells with a C-terminal His Tag.
Catalog No. 50-208-5266