Learn More
Creative Biomart Recombinant Human ERBB2 protein, His-tag, GMP Grade
Supplier:  Creative Biomart ERBB2111THP
Encompass_Preferred
Recombinant Human ERBB2 protein GMP Grade(NP_001005862)(630 aa) was expressed in HEK293 cells with a C-terminal His Tag.
Catalog No. 50-208-5299