Learn More
Apexbio Technology LLC Recombinant Human Erythropoietin/EPO 100ug
SDP
Catalog No. 5000094855
Encompass_Preferred
Recombinant Human Erythropoietin EPO is a glycoprotein hormone targeting the erythropoietin receptor (EPO R) It is designed to stimulate erythroid progenitor cell survival and differentiation thereby regulating erythropoiesis Recombinant Human Erythropoietin EPO exerts its biological activity primarily through binding and dimerization of EPO R followed by activation of the Janus kinase 2 (JAK2)-mediated signaling pathway Based on these pharmacological properties Recombinant Human Erythropoietin EPO holds research potential in studies of erythropoiesis hematopoietic disorders tissue protection mechanisms and hypoxia-associated signaling pathways
Catalog No. 50-000-94855 Supplier Apexbio Technology LLC Supplier No. PH20260.1
Recombinant Human Erythropoietin EPO is a glycoprotein hormone targeting the erythropoietin receptor (EPO R) It is designed to stimulate erythroid progenitor cell survival and differentiation thereby regulating erythropoiesis Recombinant Human Erythropoietin EPO exerts its biological activity primarily through binding and dimerization of EPO R followed by activation of the Janus kinase 2 (JAK2)-mediated signaling pathway Based on these pharmacological properties Recombinant Human Erythropoietin EPO holds research potential in studies of erythropoiesis hematopoietic disorders tissue protection mechanisms and hypoxia-associated signaling pathways