Learn More
Abcam Recombinant Human EXOSC8 protein, 50UG
Supplier:  Abcam AB18027850UG
Encompass_Preferred
Recombinant Human EXOSC8 protein is a Human Full Length protein, in the 1 to 276 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE, MS.
Protein/ Peptide/ Enzyme Synonyms: OIP2, RRP43, EXOSC8, Exosome complex component RRP43, Exosome component 8, Opa-interacting protein 2, Ribosomal RNA-processing protein 43, p9, OIP-2
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-7997