Learn More
R&D Systems™ Recombinant Human FABP9/T-FABP Protein
Description
Specifications
Specifications
| Accession Number | Q0Z7S8 |
| Characterization | Bioactivity not tested |
| For Use With (Application) | Flow Cytometry |
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS, EDTA and DTT. |
| Gene ID (Entrez) | 646480 |
| Molecular Weight (g/mol) | 16kDa |
| Quantity | 100 μg |
| Source | E. coli-derived human FABP9/T-FABP protein Met1-Val132,with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10EU per 1μg of the protein by the LAL method. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.