Learn More
ACROBiosystems Recombinant Protein;Human Fc gamma RIIA / CD32a (R167) Protein (SPR verified);HEK293;100UG;CDA,CD32a,FCGR2A,CD32,FCG2 ,FCGR2A1,IGFR2,Fcr,Fcgr,Fc receptor,Fc gamma receptor
Catalog No. NC0633178
Encompass_Preferred
Recombinant Protein;Human Fc gamma RIIA / CD32a (R167) Protein (SPR verified);100UG;Human CD32a (R167) (SPR verified) (CDA-H5221) is expressed from human 293 cells (HEK293). It contains AA Ala 36 - Ile 218 (Accession # P12318-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 21.2 kDa. The protein migrates as 25-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD32a (R167) (Cat. No. CDA-H5221) on CM5 Chip via anti-His antibody, can bind  Rituximab with an affinity constant of 3.12 μM as determined in a SPR assay (Biacore T200) (QC tested).
Catalog No. NC0633178 Supplier ACROBiosystems Supplier No. CDAH5221100UG
Recombinant Protein;Human Fc gamma RIIA / CD32a (R167) Protein (SPR verified);100UG;Human CD32a (R167) (SPR verified) (CDA-H5221) is expressed from human 293 cells (HEK293). It contains AA Ala 36 - Ile 218 (Accession # P12318-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 21.2 kDa. The protein migrates as 25-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD32a (R167) (Cat. No. CDA-H5221) on CM5 Chip via anti-His antibody, can bind  Rituximab with an affinity constant of 3.12 μM as determined in a SPR assay (Biacore T200) (QC tested).