Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Fc gamma RIIA/CD32a (R167) Protein

Catalog No. 1330CD01MCF
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-30C-D01MCF 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 13-30C-D01MCF Supplier R&D Systems™ Supplier No. 1330CD01M/CF
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity

The Recombinant Human Fc gamma RIIA/CD32a (R167) Protein is derived from NS0. The Recombinant Human Fc gamma RIIA/CD32a (R167) Protein has been validated for the following applications: Binding Activity.

Order Info

Ships at ambient temperature.

Specifications

Accession Number AAA35827
For Use With (Application) Binding Activity
Formulation Lyophilized from a 0.2μm filtered solution in PBS
Gene ID (Entrez) 2212
Molecular Weight (g/mol) Observed M.W.: 26-36 kDa, under reducing conditions.
Quantity 1 mg
Source Mouse myeloma cell line,NS0-derived human Fc gamma RIIA/CD32a protein Ala36-Ile218,with a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method
Gene Alias CD32 antigen, CD32A, CD32MGC23887
Gene Symbol FCGR2A
Biological Activity <80nM
Product Type Recombinant protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.