Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human FGF-9, Animal-Free Protein

Catalog No. QK0390250
Change view
Click to view available options
Quantity:
250 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
QK0390250 250 μg
QK0390050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. QK0390250 Supplier R&D Systems™ Supplier No. QK0390250
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
No significant difference between EC50 of reference and test lots

Specifications

Accession Number P31371.3
For Use With (Application) Bioactivity
Formulation Lyophilized from HEPES/NaCl
Gene ID (Entrez) 2254
Molecular Weight (g/mol) M.W.-observed: Monomeric FGF-9 protein only, M.W.-theroretical: 23 kDa
Quantity 250 μg
Source E. coli-derived human FGF-9 protein
Storage Requirements Store lyophilized protein between -20 and -80 °C until the date of expiry. Avoid freeze-thaw cycles.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias FGF9, FGF-9, Fibroblast growth factor 9, fibroblast growth factor 9 (glia-activating factor), GAF, glia-activating factor, HBFG-9, HBGF-9, Heparin-binding growth factor 9, MGC119914, MGC119915, SYNS3
Conjugate Unconjugated
Reconstitution Resuspend in water at >100 μg/ml, prepare single use aliquots, add carrier protein if desired.
Protein FGF-9
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.