Learn More
R&D Systems™ Recombinant Human FGF acidic/FGF1 (aa 16-155) Protein
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 and EDTA. |
| Gene ID (Entrez) | 2246 |
| Molecular Weight (g/mol) | 15.5 kDa |
| Name | FGF acidic aa 16-155 |
| Quantity | 25 μg |
| Source | E. coli-derived human FGF acidic/FGF1 protein Phe16-Asp155, with an N-terminal Met |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, ≤ -20°C under sterile conditions after reconstitution. |
| Gene Alias | AFGF, alpha, alpha-ECGF, beta-ECGF, ECGF, ECGFB, ECGF-betaAcidic fibroblast growth factor, endothelial cell growth factor, beta, FGF acidic, FGF-1, FGFABeta-endothelial cell growth factor, FGF-alpha, fibroblast growth factor 1 (acidic), GLIO703, HBGF1, HBGF-1, heparin-binding growth factor 1 |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.